Product Details

SNP ID
rs6076347
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:25418154 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCACTCAGATGGGAATATGGTAGC[A/G]TCTTGCCAAATGCATTACGATTTCA
Phenotype
MIM: 610608
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GINS1 PubMed Links

Gene Details

Gene
GINS1
Gene Name
GINS complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021067.4 384 Missense Mutation ATC,GTC I97V NP_066545.3
XM_017028162.1 384 Missense Mutation ATC,GTC I62V XP_016883651.1

View Full Product Details