Product Details
- SNP ID
-
rs11764733
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:151379575 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCACTCACCGTGTCTCTCAGGCT[C/G]TCAGAAACCAGAGCGAAAATGGACA
- Phenotype
-
MIM: 607981
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NUB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115790539] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NUB1
- Gene Name
- negative regulator of ubiquitin like proteins 1
There are no transcripts associated with this gene.
- Gene
- WDR86
- Gene Name
- WD repeat domain 86
View Full Product Details