Product Details

SNP ID
rs34783816
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:36099653 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCTGCAATAGTAAATCATTCTTTCT[A/T]CTCTTTTGAACTAAGTACAGTAAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LMBRD2 PubMed Links

Gene Details

Gene
LMBRD2
Gene Name
LMBR1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007527.1 7014 Intron NP_001007528.1
XM_011514162.2 7014 Intron XP_011512464.1
XM_017010024.1 7014 UTR 3 XP_016865513.1

View Full Product Details