Product Details

SNP ID
rs10407394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14693371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTCCTGTAAGGGTTTTGATTGTG[A/G]CCACTCTGCTGGAGATCCCCCAAAA
Phenotype
MIM: 611811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF333 PubMed Links

Gene Details

Gene
ZNF333
Gene Name
zinc finger protein 333
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300912.1 Intron NP_001287841.1
NM_032433.2 Intron NP_115809.1
XM_005260098.3 Intron XP_005260155.1
XM_006722924.2 Intron XP_006722987.1
XM_011528362.2 Intron XP_011526664.1
XM_011528363.2 Intron XP_011526665.1
XM_011528365.2 Intron XP_011526667.1
XM_011528366.2 Intron XP_011526668.1
XM_011528367.2 Intron XP_011526669.1
XM_011528368.2 Intron XP_011526670.1
XM_011528369.2 Intron XP_011526671.1
XM_011528371.2 Intron XP_011526673.1
XM_011528372.2 Intron XP_011526674.1
XM_017027367.1 Intron XP_016882856.1
XM_017027368.1 Intron XP_016882857.1
XM_017027369.1 Intron XP_016882858.1
XM_017027370.1 Intron XP_016882859.1
XM_017027371.1 Intron XP_016882860.1

View Full Product Details