Product Details

SNP ID
rs3762735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101564948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTATCCTAAAAATGAGAGTACAC[C/G]CCCTTCTGAAGAGCTAGAGTTGGAT
Phenotype
MIM: 615210 MIM: 615423
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PCNP PubMed Links

Gene Details

Gene
PCNP
Gene Name
PEST proteolytic signal containing nuclear protein
There are no transcripts associated with this gene.

Gene
TRMT10C
Gene Name
tRNA methyltransferase 10C, mitochondrial RNase P subunit
There are no transcripts associated with this gene.

View Full Product Details