Product Details

SNP ID
rs1054713
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:50820245 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGGCAACTCCACGACCTTCACAGC[A/G]TCTGTGATGGTATCAGCAGGCTCTG
Phenotype
MIM: 147910 MIM: 610601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK1 PubMed Links

Gene Details

Gene
KLK1
Gene Name
kallikrein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002257.3 1340 Silent Mutation GAC,GAT D135D NP_002248.1
XM_011526942.1 1340 Silent Mutation GAC,GAT D33D XP_011525244.1
Gene
KLK15
Gene Name
kallikrein related peptidase 15
There are no transcripts associated with this gene.

Gene
MGC45922
Gene Name
uncharacterized LOC284365
There are no transcripts associated with this gene.

View Full Product Details