Product Details

SNP ID
rs249355
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:180908671 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CATTCTCCTGTTTCCTGTCTCCTAA[A/G]ACCAATGCAGAGGCCATGGAAGTGC
Phenotype
MIM: 615606
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTNL8 PubMed Links

Gene Details

Gene
BTNL8
Gene Name
butyrophilin like 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040462.2 174 Silent Mutation AAA,AAG K45K NP_001035552.1
NM_001159707.1 174 Intron NP_001153179.1
NM_001159708.1 174 Silent Mutation AAA,AAG K45K NP_001153180.1
NM_001159709.1 174 Intron NP_001153181.1
NM_001159710.1 174 Intron NP_001153182.1
NM_024850.2 174 Silent Mutation AAA,AAG K45K NP_079126.1
XM_011534649.1 174 Silent Mutation AAA,AAG K45K XP_011532951.1
XM_011534650.2 174 UTR 5 XP_011532952.1
XM_011534651.2 174 Intron XP_011532953.1
XM_011534652.2 174 Intron XP_011532954.1
XM_011534653.2 174 Intron XP_011532955.1

View Full Product Details