Product Details
- SNP ID
-
rs2684339
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52691478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGTGCACACGGGGTCAGAGGGAC[A/C]GGGCAAGGCCCCCACCTGCACCCGC
- Phenotype
-
MIM: 611158
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT77
PubMed Links
Gene Details
- Gene
- KRT77
- Gene Name
- keratin 77
View Full Product Details