Product Details

SNP ID
rs692898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68891935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCATCCCTCAGGATGCCCACT[A/G]TGGGGGATGTCCTCAGTGCAGACCT
Phenotype
MIM: 611834
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL21 PubMed Links

Gene Details

Gene
MRPL21
Gene Name
mitochondrial ribosomal protein L21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181514.1 Intron NP_852615.1
NM_181515.1 Intron NP_852616.1
XM_005273823.4 Intron XP_005273880.1

View Full Product Details