Product Details

SNP ID
rs12814230
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:27765511 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGTGGCCAGATCTGTTTGCTTACT[C/G]TCCTGTGGCATAATATAGAACTCAT
Phenotype
MIM: 611995
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MANSC4 PubMed Links

Gene Details

Gene
MANSC4
Gene Name
MANSC domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146221.1 Intron NP_001139693.1
XM_006719024.3 Intron XP_006719087.1
XM_011520542.2 Intron XP_011518844.1
Gene
MRPS35
Gene Name
mitochondrial ribosomal protein S35
There are no transcripts associated with this gene.

View Full Product Details