Product Details

SNP ID
rs1043846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:80272335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGACAAGTGGGGCAGGTAGGG[C/G]AGGTGCTGCTGGCCATGGATGGAAA
Phenotype
MIM: 610550
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAT1A PubMed Links

Gene Details

Gene
MAT1A
Gene Name
methionine adenosyltransferase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000429.2 2889 UTR 3 NP_000420.1
XM_005269842.4 2889 UTR 3 XP_005269899.1
XM_005269843.4 2889 UTR 3 XP_005269900.1

View Full Product Details