Product Details

SNP ID
rs2920
Assay Type
Functionally tested
NCBI dbSNP Submissions
68
Location
Chr.1:23558289 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCTTCGCTCCTGAGCACCAGGTT[C/T]AGTCTCCAGGAAGGGATTTGGTGAA
Phenotype
MIM: 600277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ID3 PubMed Links

Gene Details

Gene
ID3
Gene Name
inhibitor of DNA binding 3, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002167.4 880 UTR 3 NP_002158.3

View Full Product Details