Product Details

SNP ID
rs852441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5621055 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAGGTATGGGCGCCAGCAAGGAC[A/G]TAGCAGCAGGCTGCCCCCAAGCCCG
Phenotype
MIM: 609948
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107986762 PubMed Links

Gene Details

Gene
LOC107986762
Gene Name
uncharacterized LOC107986762
There are no transcripts associated with this gene.

Gene
RNF216
Gene Name
ring finger protein 216
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207111.3 4575 UTR 3 NP_996994.1
NM_207116.2 4575 UTR 3 NP_996999.1
XM_005249785.2 4575 UTR 3 XP_005249842.1
XM_011515436.1 4575 UTR 3 XP_011513738.1
XM_017012363.1 4575 UTR 3 XP_016867852.1
XM_017012364.1 4575 Intron XP_016867853.1
XM_017012365.1 4575 UTR 3 XP_016867854.1

View Full Product Details