Product Details

SNP ID
rs700240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:39374813 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCCAAGGGTATATTACTATTTTT[G/T]GATGTGTTTAGGAATTCTATTCTTA
Phenotype
MIM: 601236
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DAB2 PubMed Links

Gene Details

Gene
DAB2
Gene Name
DAB2, clathrin adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244871.1 Intron NP_001231800.1
NM_001343.3 Intron NP_001334.2

View Full Product Details