Product Details

SNP ID
rs1303567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38477514 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCATCTGGTACATAGGTGAATAC[A/C]TTCCTTTTTTGCCAGCTTTATTGAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RALGAPB PubMed Links

Gene Details

Gene
RALGAPB
Gene Name
Ral GTPase activating protein non-catalytic beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282917.1 Intron NP_001269846.1
NM_001282918.1 Intron NP_001269847.1
NM_020336.3 Intron NP_065069.1
XM_005260462.2 Intron XP_005260519.1
XM_005260463.2 Intron XP_005260520.1
XM_005260464.2 Intron XP_005260521.1
XM_005260465.2 Intron XP_005260522.1
XM_017027966.1 Intron XP_016883455.1
XM_017027967.1 Intron XP_016883456.1
XM_017027968.1 Intron XP_016883457.1
XM_017027969.1 Intron XP_016883458.1
XM_017027970.1 Intron XP_016883459.1

View Full Product Details