Product Details

SNP ID
rs1389499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93574167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAACAAGTACATAGCAGTGCTTT[C/T]TACATAGAAGGCTGTTCATAGCAGG
Phenotype
MIM: 605117
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOCS2 PubMed Links

Gene Details

Gene
SOCS2
Gene Name
suppressor of cytokine signaling 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270467.1 Intron NP_001257396.1
NM_001270468.1 Intron NP_001257397.1
NM_001270469.1 Intron NP_001257398.1
NM_001270470.1 Intron NP_001257399.1
NM_001270471.1 Intron NP_001257400.1
NM_003877.4 Intron NP_003868.1
XM_011538929.1 Intron XP_011537231.1
XM_011538935.1 Intron XP_011537237.1
XM_011538936.1 Intron XP_011537238.1
XM_017020147.1 Intron XP_016875636.1
XM_017020148.1 Intron XP_016875637.1
XM_017020149.1 Intron XP_016875638.1
XM_017020150.1 Intron XP_016875639.1
XM_017020151.1 Intron XP_016875640.1
XM_017020152.1 Intron XP_016875641.1
XM_017020153.1 Intron XP_016875642.1
XM_017020154.1 Intron XP_016875643.1
XM_017020155.1 Intron XP_016875644.1
XM_017020156.1 Intron XP_016875645.1
Gene
SOCS2-AS1
Gene Name
SOCS2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details