Product Details

SNP ID
rs12610267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38254093 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAACAGAGGGACTGCATGTCACC[A/G]TGGGAGAAGGGCCATGCCTTCCACC
Phenotype
MIM: 608153
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R14A PubMed Links

Gene Details

Gene
PPP1R14A
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243947.1 Intron NP_001230876.1
NM_033256.2 Intron NP_150281.1

View Full Product Details