Product Details

SNP ID
rs1251074
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:75725005 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGACGGAACGTAGCCTAGCGTTT[A/C]ATTTTCCGTATCCTCCCGTCAGGCG
Phenotype
MIM: 607008
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACADM PubMed Links

Gene Details

Gene
ACADM
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000016.5 Intron NP_000007.1
NM_001127328.2 Intron NP_001120800.1
NM_001286042.1 Intron NP_001272971.1
NM_001286043.1 Intron NP_001272972.1
NM_001286044.1 Intron NP_001272973.1
Gene
SLC44A5
Gene Name
solute carrier family 44 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130058.1 Intron NP_001123530.1
NM_001320283.1 Intron NP_001307212.1
NM_001320285.1 Intron NP_001307214.1
NM_001320287.1 Intron NP_001307216.1
NM_152697.4 Intron NP_689910.2
XM_006710445.3 Intron XP_006710508.1
XM_011540986.2 Intron XP_011539288.1
XM_011540987.2 Intron XP_011539289.1
XM_017000607.1 Intron XP_016856096.1
XM_017000608.1 Intron XP_016856097.1
XM_017000609.1 Intron XP_016856098.1
XM_017000610.1 Intron XP_016856099.1
XM_017000611.1 Intron XP_016856100.1

View Full Product Details