Product Details

SNP ID
rs920236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:165929602 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGCAGCCCCATGTGTGCAGGGCC[A/T]TCAGTGCTGCTGGAAAATCATGAAG
Phenotype
MIM: 610652
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00473 PubMed Links

Gene Details

Gene
LINC00473
Gene Name
long intergenic non-protein coding RNA 473
There are no transcripts associated with this gene.

Gene
PDE10A
Gene Name
phosphodiesterase 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130690.2 Intron NP_001124162.1
NM_006661.3 Intron NP_006652.1
XM_006715321.3 Intron XP_006715384.1
XM_011535387.2 Intron XP_011533689.2
XM_011535388.2 Intron XP_011533690.1
XM_011535393.2 Intron XP_011533695.1
XM_017010194.1 Intron XP_016865683.1
XM_017010195.1 Intron XP_016865684.1
XM_017010196.1 Intron XP_016865685.1
XM_017010197.1 Intron XP_016865686.1

View Full Product Details