Product Details

SNP ID
rs1042715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:65824566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCCCTCCGACTCTCCGGTGCCG[C/G]CGCTGCCTGCTCCCGCCACCCTAGG
Phenotype
MIM: 600698
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HMGA2 PubMed Links

Gene Details

Gene
HMGA2
Gene Name
high mobility group AT-hook 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300918.1 107 UTR 5 NP_001287847.1
NM_001300919.1 107 UTR 5 NP_001287848.1
NM_003483.4 107 UTR 5 NP_003474.1
NM_003484.1 107 UTR 5 NP_003475.1
XM_017019989.1 107 Intron XP_016875478.1
Gene
RPSAP52
Gene Name
ribosomal protein SA pseudogene 52
There are no transcripts associated with this gene.

View Full Product Details