Product Details

SNP ID
rs13400
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:57757610 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGCAGTCCGGATCCGGAGGAACTC[C/G]TCTTTGTCCCTGGTAGGAGAGACAC
Phenotype
MIM: 602701
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLMAP PubMed Links

Gene Details

Gene
SLMAP
Gene Name
sarcolemma associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304420.2 1185 UTR 5 NP_001291349.1
NM_001304421.2 1185 UTR 5 NP_001291350.1
NM_001304422.2 1185 Intron NP_001291351.1
NM_001304423.2 1185 Intron NP_001291352.1
NM_001311178.1 1185 Intron NP_001298107.1
NM_001311179.1 1185 Intron NP_001298108.1
NM_007159.4 1185 UTR 5 NP_009090.2
XM_005265456.3 1185 UTR 5 XP_005265513.1
XM_005265457.3 1185 UTR 5 XP_005265514.1
XM_005265458.3 1185 UTR 5 XP_005265515.1
XM_005265459.3 1185 UTR 5 XP_005265516.1
XM_005265460.3 1185 UTR 5 XP_005265517.1
XM_005265461.3 1185 UTR 5 XP_005265518.1
XM_005265462.3 1185 UTR 5 XP_005265519.1
XM_005265464.3 1185 UTR 5 XP_005265521.1
XM_005265465.3 1185 UTR 5 XP_005265522.1
XM_005265466.3 1185 UTR 5 XP_005265523.1
XM_005265467.3 1185 UTR 5 XP_005265524.1
XM_005265468.3 1185 UTR 5 XP_005265525.1
XM_005265470.3 1185 UTR 5 XP_005265527.1
XM_005265471.3 1185 UTR 5 XP_005265528.1
XM_005265472.3 1185 UTR 5 XP_005265529.1
XM_006713326.2 1185 UTR 5 XP_006713389.1
XM_011534103.1 1185 UTR 5 XP_011532405.1
XM_011534104.1 1185 UTR 5 XP_011532406.1
XM_011534105.1 1185 UTR 5 XP_011532407.1
XM_011534106.1 1185 Intron XP_011532408.1
XM_017007146.1 1185 UTR 5 XP_016862635.1
XM_017007147.1 1185 UTR 5 XP_016862636.1
XM_017007148.1 1185 UTR 5 XP_016862637.1
XM_017007149.1 1185 UTR 5 XP_016862638.1
XM_017007150.1 1185 UTR 5 XP_016862639.1
XM_017007151.1 1185 UTR 5 XP_016862640.1
XM_017007152.1 1185 UTR 5 XP_016862641.1
XM_017007153.1 1185 UTR 5 XP_016862642.1
XM_017007154.1 1185 UTR 5 XP_016862643.1
XM_017007155.1 1185 UTR 5 XP_016862644.1
XM_017007156.1 1185 UTR 5 XP_016862645.1
XM_017007157.1 1185 UTR 5 XP_016862646.1
XM_017007158.1 1185 UTR 5 XP_016862647.1
XM_017007159.1 1185 UTR 5 XP_016862648.1
XM_017007160.1 1185 UTR 5 XP_016862649.1
XM_017007161.1 1185 UTR 5 XP_016862650.1
XM_017007162.1 1185 UTR 5 XP_016862651.1
XM_017007163.1 1185 UTR 5 XP_016862652.1
XM_017007164.1 1185 UTR 5 XP_016862653.1
XM_017007165.1 1185 UTR 5 XP_016862654.1
XM_017007166.1 1185 UTR 5 XP_016862655.1
XM_017007167.1 1185 UTR 5 XP_016862656.1
XM_017007168.1 1185 UTR 5 XP_016862657.1
XM_017007169.1 1185 UTR 5 XP_016862658.1
XM_017007170.1 1185 Intron XP_016862659.1

View Full Product Details