Product Details
- SNP ID
-
rs777513
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:126481303 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAAGGCTGGAGAATCAGGTGGTTTT[A/T]TTTACCCTATATTGCTGCCCACACA
- Phenotype
-
MIM: 613012
MIM: 615746
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UROC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7620946] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UROC1
- Gene Name
- urocanate hydratase 1
- Gene
- ZXDC
- Gene Name
- ZXD family zinc finger C
There are no transcripts associated with this gene.
View Full Product Details