Product Details

SNP ID
rs916418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:49775557 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCCCACGGCCCCCAACCACCCCA[A/G]CCGGTCCCCGGAGTCTAAGGCCTCT
Phenotype
MIM: 604589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD1 PubMed Links

Gene Details

Gene
BRD1
Gene Name
bromodomain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304808.1 Intron NP_001291737.1
NM_001304809.1 Intron NP_001291738.1
XM_011530073.1 Intron XP_011528375.1
XM_017028714.1 Intron XP_016884203.1
XM_017028715.1 Intron XP_016884204.1
XM_017028716.1 Intron XP_016884205.1
XM_017028717.1 Intron XP_016884206.1
XM_017028718.1 Intron XP_016884207.1
XM_017028719.1 Intron XP_016884208.1
XM_017028720.1 Intron XP_016884209.1
XM_017028721.1 Intron XP_016884210.1
XM_017028722.1 Intron XP_016884211.1
XM_017028723.1 Intron XP_016884212.1

View Full Product Details