Product Details
- SNP ID
-
rs1279195
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:109548660 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCAATAGACGGTGCCTCAGCCTGC[C/T]GAGCCGCAGTTTCCGTGGTGTGAGT
- Phenotype
-
MIM: 139370
MIM: 606916
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GNAI3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144431312] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNAI3
- Gene Name
- G protein subunit alpha i3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006496.3 |
97 |
UTR 5 |
|
|
NP_006487.1 |
- Gene
- GPR61
- Gene Name
- G protein-coupled receptor 61
There are no transcripts associated with this gene.
View Full Product Details