Product Details

SNP ID
hCV8856726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:241536535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACAGCTAAAGCTACTCAAGAC[A/C]CCATTTTTTGACTGAAGCCAAGAGC
Phenotype
MIM: 603538
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KMO PubMed Links

Gene Details

Gene
KMO
Gene Name
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003679.4 Intron NP_003670.2

View Full Product Details