Product Details

SNP ID
rs10310
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:141318833 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGATGAACACCTAGCAAAACAATA[C/T]AAATTTGCTGAAAAACAAAATTACA
Phenotype
MIM: 600573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF7 PubMed Links

Gene Details

Gene
TAF7
Gene Name
TATA-box binding protein associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005642.2 1952 UTR 3 NP_005633.2

View Full Product Details