Product Details

SNP ID
rs1803055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:10637742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCACACTTGCTGTTGAATAACAA[C/G]GGAATCAAGGCTCCCCTAGATGGGG
Phenotype
MIM: 601920 MIM: 615958
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
JAG1 PubMed Links

Gene Details

Gene
JAG1
Gene Name
jagged 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000214.2 5929 UTR 3 NP_000205.1
XM_017027707.1 5929 UTR 3 XP_016883196.1
Gene
SLX4IP
Gene Name
SLX4 interacting protein
There are no transcripts associated with this gene.

View Full Product Details