Product Details

SNP ID
rs1274318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:67245347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCTTTAGATTTAGCCATTTTA[A/G]TAGTATCTCGTTGGGATTTTTCTTT
Phenotype
MIM: 603930 MIM: 606958
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPHN PubMed Links

Gene Details

Gene
GPHN
Gene Name
gephyrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024218.1 Intron NP_001019389.1
NM_020806.4 Intron NP_065857.1
XM_005267254.3 Intron XP_005267311.1
XM_011536340.2 Intron XP_011534642.1
XM_011536342.2 Intron XP_011534644.1
XM_011536343.2 Intron XP_011534645.1
XM_011536344.2 Intron XP_011534646.1
XM_011536345.2 Intron XP_011534647.1
XM_011536346.2 Intron XP_011534648.1
XM_011536347.2 Intron XP_011534649.1
XM_017020913.1 Intron XP_016876402.1
XM_017020914.1 Intron XP_016876403.1
XM_017020915.1 Intron XP_016876404.1
XM_017020916.1 Intron XP_016876405.1
XM_017020917.1 Intron XP_016876406.1
XM_017020918.1 Intron XP_016876407.1
XM_017020919.1 Intron XP_016876408.1
XM_017020920.1 Intron XP_016876409.1
XM_017020921.1 Intron XP_016876410.1
XM_017020922.1 Intron XP_016876411.1
XM_017020923.1 Intron XP_016876412.1
XM_017020924.1 Intron XP_016876413.1
XM_017020925.1 Intron XP_016876414.1
XM_017020926.1 Intron XP_016876415.1
Gene
MPP5
Gene Name
membrane palmitoylated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256550.1 Intron NP_001243479.1
NM_022474.3 Intron NP_071919.2
XM_005268003.1 Intron XP_005268060.1
XM_011537086.2 Intron XP_011535388.1
XM_011537087.2 Intron XP_011535389.1

View Full Product Details