Product Details

SNP ID
rs950172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64847777 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGTCCTTGGAACCCTCACCAGAT[A/G]TGGTGGCCACCCTAAGGTCCTTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHO2 PubMed Links

Gene Details

Gene
PLEKHO2
Gene Name
pleckstrin homology domain containing O2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195059.1 Intron NP_001181988.1
NM_025201.4 Intron NP_079477.2

View Full Product Details