Product Details

SNP ID
rs938951
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:65196071 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTACACTGGGTTTCTAGCAAGAACA[A/G]AAGCGAATGCAAAGGGAAATGTACA
Phenotype
MIM: 603489
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CILP PubMed Links

Gene Details

Gene
CILP
Gene Name
cartilage intermediate layer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003613.3 4396 UTR 3 NP_003604.3
XM_017022678.1 4396 Intron XP_016878167.1
XM_017022679.1 4396 Intron XP_016878168.1

View Full Product Details