Product Details
- SNP ID
-
rs5174
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
82
- Location
-
Chr.1:53247055 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATCATCTTCAAGGCTTAATGCCACT[C/T]GCTGGGGAGACAAACCAAAGAATTC
- Phenotype
-
MIM: 602600
MIM: 602603
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100507564
PubMed Links
Gene Details
- Gene
- LOC100507564
- Gene Name
- uncharacterized LOC100507564
There are no transcripts associated with this gene.
- Gene
- LRP8
- Gene Name
- LDL receptor related protein 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001018054.2 |
2649 |
Missense Mutation |
CAA,CGA |
Q893R |
NP_001018064.1 |
NM_004631.4 |
2649 |
Missense Mutation |
CAA,CGA |
Q952R |
NP_004622.2 |
NM_017522.4 |
2649 |
Missense Mutation |
CAA,CGA |
Q689R |
NP_059992.3 |
NM_033300.3 |
2649 |
Missense Mutation |
CAA,CGA |
Q782R |
NP_150643.2 |
XM_005271173.3 |
2649 |
Missense Mutation |
CAA,CGA |
Q965R |
XP_005271230.1 |
XM_005271174.3 |
2649 |
Missense Mutation |
CAA,CGA |
Q836R |
XP_005271231.1 |
XM_005271175.3 |
2649 |
Missense Mutation |
CAA,CGA |
Q823R |
XP_005271232.1 |
XM_006710881.3 |
2649 |
Missense Mutation |
CAA,CGA |
Q906R |
XP_006710944.1 |
XM_006710882.3 |
2649 |
Missense Mutation |
CAA,CGA |
Q890R |
XP_006710945.1 |
XM_011542094.2 |
2649 |
Missense Mutation |
CAA,CGA |
Q923R |
XP_011540396.1 |
XM_011542095.2 |
2649 |
Missense Mutation |
CAA,CGA |
Q793R |
XP_011540397.1 |
XM_011542096.2 |
2649 |
Intron |
|
|
XP_011540398.1 |
XM_017002265.1 |
2649 |
Missense Mutation |
CAA,CGA |
Q910R |
XP_016857754.1 |
XM_017002266.1 |
2649 |
Missense Mutation |
CAA,CGA |
Q831R |
XP_016857755.1 |
XM_017002267.1 |
2649 |
Missense Mutation |
CAA,CGA |
Q546R |
XP_016857756.1 |
XM_017002268.1 |
2649 |
Missense Mutation |
CAA,CGA |
Q546R |
XP_016857757.1 |
- Gene
- MAGOH
- Gene Name
- mago homolog, exon junction complex core component
There are no transcripts associated with this gene.
View Full Product Details