Product Details
- SNP ID
-
rs11076829
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2787602 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGGGGCTTAAAGTGCCAGGAGAG[C/T]GCGAGGCTCAGCCAAGGAGTAATGA
- Phenotype
-
MIM: 613797
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS33
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80014552] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRSS33
- Gene Name
- protease, serine 33
- Gene
- PRSS41
- Gene Name
- protease, serine 41
There are no transcripts associated with this gene.
View Full Product Details