Product Details

SNP ID
rs4807322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2756702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAACTCAGGGGAGCCTCCCACCCC[C/T]GGGAAGCCGGTGTCTGCTGGAGACG
Phenotype
MIM: 603419
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGTA PubMed Links

Gene Details

Gene
SGTA
Gene Name
small glutamine rich tetratricopeptide repeat containing alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003021.3 Intron NP_003012.1
XM_011528178.2 Intron XP_011526480.1

View Full Product Details