Product Details

SNP ID
rs4843748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87959323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTAAAATGTAAAACCCATTCCTG[A/G]CTCACAGGCTGCATAGAAGCAGGTG
Phenotype
MIM: 611564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BANP PubMed Links

Gene Details

Gene
BANP
Gene Name
BTG3 associated nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173539.1 636 Intron NP_001167010.1
NM_001173540.1 636 Intron NP_001167011.1
NM_001173541.1 636 Intron NP_001167012.1
NM_001173542.1 636 Intron NP_001167013.1
NM_001173543.1 636 Intron NP_001167014.1
NM_017869.3 636 Intron NP_060339.2
NM_079837.2 636 Intron NP_524576.2
XM_005256008.3 636 Intron XP_005256065.1
XM_005256009.3 636 Intron XP_005256066.1
XM_011523170.2 636 Intron XP_011521472.1
XM_011523172.2 636 Intron XP_011521474.1
XM_011523174.2 636 Intron XP_011521476.1
XM_011523175.2 636 Intron XP_011521477.1
XM_011523176.2 636 Intron XP_011521478.1
XM_011523177.2 636 UTR 5 XP_011521479.1
XM_011523178.2 636 Intron XP_011521480.1
XM_011523179.2 636 Intron XP_011521481.1
XM_011523180.2 636 Intron XP_011521482.1
XM_011523181.2 636 Intron XP_011521483.1
XM_011523182.1 636 Intron XP_011521484.1
XM_011523183.2 636 Intron XP_011521485.1
XM_011523184.2 636 Intron XP_011521486.1
XM_011523186.2 636 Intron XP_011521488.1
XM_017023378.1 636 Intron XP_016878867.1
XM_017023379.1 636 Intron XP_016878868.1
XM_017023380.1 636 Intron XP_016878869.1
XM_017023381.1 636 Intron XP_016878870.1
XM_017023382.1 636 Intron XP_016878871.1
XM_017023383.1 636 Intron XP_016878872.1

View Full Product Details