Product Details
- SNP ID
-
rs600295
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:114177038 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGCTTCCTGTGGCCAAAGCTGGAA[A/G]AATCTGAGCAAGGAAATACAGTAGT
- Phenotype
-
MIM: 605796
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC186
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77689974] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC186
- Gene Name
- coiled-coil domain containing 186
There are no transcripts associated with this gene.
- Gene
- MIR2110
- Gene Name
- microRNA 2110
There are no transcripts associated with this gene.
- Gene
- TDRD1
- Gene Name
- tudor domain containing 1
View Full Product Details