Product Details

SNP ID
rs546183
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:183188618 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTCAACCTTGTAGAGTTTTTGAA[A/G]GGATGAACAATTACTGTAAAGTGCC
Phenotype
MIM: 150292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMC2 PubMed Links

Gene Details

Gene
LAMC2
Gene Name
laminin subunit gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005562.2 Intron NP_005553.2
NM_018891.2 Intron NP_061486.2
XM_017001273.1 Intron XP_016856762.1

View Full Product Details