Product Details
- SNP ID
-
rs736926
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:801381 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCAAGCCGGCCTGTGCCGCCTGGG[T/C]AGGCATGGAGTTTGCTGCCGCAGAC
- Phenotype
-
MIM: 600693
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4745
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113232514] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR4745
- Gene Name
- microRNA 4745
There are no transcripts associated with this gene.
- Gene
- PTBP1
- Gene Name
- polypyrimidine tract binding protein 1
View Full Product Details