Product Details

SNP ID
rs717343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92292241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGGGCCCCAGTCGGTAAAACT[C/T]TTTGTGAAGTCGGGTGCACCGCCCT
Phenotype
MIM: 610606 MIM: 610637
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPEB3 PubMed Links

Gene Details

Gene
CPEB3
Gene Name
cytoplasmic polyadenylation element binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178137.1 Intron NP_001171608.1
NM_014912.4 Intron NP_055727.3
Gene
MARCH5
Gene Name
membrane associated ring-CH-type finger 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017824.4 Intron NP_060294.1
XM_011539886.2 Intron XP_011538188.1

View Full Product Details