Product Details

SNP ID
rs12074893
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:40162447 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGTTTTCAAGTCTTTTACAAATA[C/T]AAATATTGGCCATTATTATTTCTGC
Phenotype
MIM: 180610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RLF PubMed Links

Gene Details

Gene
RLF
Gene Name
rearranged L-myc fusion
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012421.3 Intron NP_036553.2
XM_017002016.1 Intron XP_016857505.1
XM_017002017.1 Intron XP_016857506.1
XM_017002018.1 Intron XP_016857507.1

View Full Product Details