Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
che-1
Entrez Gene ID
183847
Gene Name
protein-coding
Gene Aliases
CELE_C55B7.12
Chromosome Location
Chr.I: 6517529 - 6520918 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083128.1 NP_001076597.1 4 - 5 467 161
NM_001083129.2 NP_001076598.1 5 - 6 683 161

View Full Product Details