Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-3
Entrez Gene ID
171617
Gene Name
protein-coding
Gene Aliases
CELE_C53D5.4
Chromosome Location
Chr.I: 286040 - 289185 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373713.1 NP_001360315.1 3 - 4 453 64
NM_001373725.1 NP_001359947.1 3 - 4 453 64
NM_001381400.1 NP_001368396.1 3 - 4 303 64
NM_001381421.1 NP_001368397.1 3 - 4 303 64

View Full Product Details