Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mps-3
Entrez Gene ID
188157
Gene Name
MiRP K channel accessory Subunit
Gene Aliases
CELE_T06A4.2
Chromosome Location
Chr.I: 760848 - 761849 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058377.4 NP_490778.1 2 - 3 156 88
GenBank mRNA AY255668.1 - 2 - 3 143 88

View Full Product Details