Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exc-5
Entrez Gene ID
177692
Gene Name
protein-coding
Gene Aliases
CELE_C33D9.1
Chromosome Location
Chr.IV: 8799081 - 8813670 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027868.5 NP_001023039.1 10 - 11 1689 79
NM_001307099.3 NP_001294028.1 9 - 10 1488 79
NM_001307100.4 NP_001294029.1 11 - 12 1925 79
GenBank mRNA AB060647.1 - 10 - 11 1742 79
AF339059.1 - 10 - 11 1742 79

View Full Product Details