Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exc-5
Entrez Gene ID
177692
Gene Name
protein-coding
Gene Aliases
CELE_C33D9.1
Chromosome Location
Chr.IV: 8799081 - 8813670 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027868.5 NP_001023039.1 12 - 13 1952 71
NM_001307099.3 NP_001294028.1 11 - 12 1751 71
NM_001307100.4 NP_001294029.1 13 - 14 2188 71
GenBank mRNA AB060647.1 - 12 - 13 2005 71
AF339059.1 - 12 - 13 2005 71

View Full Product Details