Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exc-5
Entrez Gene ID
177692
Gene Name
protein-coding
Gene Aliases
CELE_C33D9.1
Chromosome Location
Chr.IV: 8799081 - 8813670 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027868.5 NP_001023039.1 6 - 7 717 109
NM_001307099.3 NP_001294028.1 5 - 6 516 109
NM_001307100.4 NP_001294029.1 7 - 8 953 109
GenBank mRNA AB060647.1 - 6 - 7 770 109
AF339059.1 - 6 - 7 770 109

View Full Product Details