Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nas-9
Entrez Gene ID
178875
Gene Name
protein-coding
Gene Aliases
CELE_C37H5.9
Chromosome Location
Chr.V: 4836027 - 4840010 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171452.2 NP_741531.1 8 - 9 1278 83

View Full Product Details