Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-17
Entrez Gene ID
190174
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_Y51H7C.2
Chromosome Location
Chr.II: 1445211 - 1446108 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061574.3 NP_493975.1 2 - 3 536 94

View Full Product Details