Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-28
Entrez Gene ID
173569
Gene Name
SKN-1 Dependent Zygotic transcript
Gene Aliases
CELE_R52.1
Chromosome Location
Chr.II: 2131070 - 2132475 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061791.4 NP_494192.1 1 - 2 125 96

View Full Product Details