Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-2
Entrez Gene ID
181993
Gene Name
C-type LECtin
Gene Aliases
CELE_B0454.7
Chromosome Location
Chr.II: 3020562 - 3023431 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062026.6 NP_494427.1 2 - 3 327 59

View Full Product Details