Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-14
Entrez Gene ID
182758
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_C18A3.8
Chromosome Location
Chr.II: 5737785 - 5738895 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062730.6 NP_495131.3 1 - 2 151 81

View Full Product Details